Learn More
Research Products International Corp Flurprimidol, 100 Milligrams
SDP
Supplier:  Research Products International Corp F200200.1
Encompass_Preferred
Synthetic inhibitor of Gibberellic acid synthesis. Also used in tissue culture to control internode elongation in liquid cultures.
Catalog No. 50-213-318