Learn More
Med Vet International Basic Equine Uterine Flush: Single Line, J1052C, Each
Catalog No. 5000495712
Encompass_Preferred
Basic Equine Uterine Flush: Single Line No Frills. • 28fr (9.3mm) o.d. • 190cm or 76" Length • On/Off slide clamp • Vented IV spike • Distal tip – Smooth open end with 4 lateral eyes • Sterile
Catalog No. 50-004-95712 Supplier Med Vet International Supplier No. JOR1052C
Basic Equine Uterine Flush: Single Line No Frills. • 28fr (9.3mm) o.d. • 190cm or 76" Length • On/Off slide clamp • Vented IV spike • Distal tip – Smooth open end with 4 lateral eyes • Sterile