Learn More
Grainger Flush Trim Profile Router Bit 1/4
Catalog No. 502766861
Encompass_Preferred
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming and shaping wood, plastic, and composite materials. It has a 1-1/4 HP motor with a variable speed range of 10,000-30,000 RPM. The router comes with a 1/4" and 1/2" collet, flush trim and round over bits, a chip deflector, and dust extraction adapter. It features soft start circuitry for smooth startups and overload protection to prevent motor burnout. The compact, ergonomic design provides excellent control and maneuverability for detail work. With its adjustable depth stop and steel baseplate, this router is well-suited for edge forming, laminate trimming, and other light routing applications.
Catalog No. 50-276-6861 Supplier Grainger Supplier No. 16Y593
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming and shaping wood, plastic, and composite materials. It has a 1-1/4 HP motor with a variable speed range of 10,000-30,000 RPM. The router comes with a 1/4" and 1/2" collet, flush trim and round over bits, a chip deflector, and dust extraction adapter. It features soft start circuitry for smooth startups and overload protection to prevent motor burnout. The compact, ergonomic design provides excellent control and maneuverability for detail work. With its adjustable depth stop and steel baseplate, this router is well-suited for edge forming, laminate trimming, and other light routing applications.