Learn More
Grainger Flush Trim Profile Router Bit 1/2
Catalog No. 502767008
Encompass_Preferred
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 27,000 RPM. The router comes with a 1/4 inch collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It is lightweight at only 4.4 lbs. This router allows for smooth trimming and profiling of countertop and cabinet edges. Its compact size makes it easy to handle and maneuver.
Catalog No. 50-276-7008 Supplier Grainger Supplier No. 16Y531
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 27,000 RPM. The router comes with a 1/4 inch collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It is lightweight at only 4.4 lbs. This router allows for smooth trimming and profiling of countertop and cabinet edges. Its compact size makes it easy to handle and maneuver.