Learn More
Grainger Flush Trim Profile Router Bit 3/8
Catalog No. 502767012
Encompass_Preferred
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 27,000 RPM. The router comes with a 1/4-inch collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It weighs 4.4 lbs. This compact router can be used for trimming plastic laminates, trimming edges of wood veneer prior to gluing, putting edges on panels, trimming Formica counter tops. It has precision ball bearing construction for smooth operation.
Catalog No. 50-276-7012 Supplier Grainger Supplier No. 16Y538
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 27,000 RPM. The router comes with a 1/4-inch collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It weighs 4.4 lbs. This compact router can be used for trimming plastic laminates, trimming edges of wood veneer prior to gluing, putting edges on panels, trimming Formica counter tops. It has precision ball bearing construction for smooth operation.