Learn More
Grainger Flush Trim Profile Router Bit 1/2
Catalog No. 502766217
Encompass_Preferred
The WESTWARD Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 24,000 RPM. The router comes with a 1/4" collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It is compact and lightweight at only 4 lbs, with soft grip handles for comfort and control. The router has a lock-on button for continuous operation. It is ideal for trimming plastic laminates, wood veneers, and solid surface materials.
Catalog No. 50-276-6217 Supplier Grainger Supplier No. 16Y549
The WESTWARD Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 24,000 RPM. The router comes with a 1/4" collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It is compact and lightweight at only 4 lbs, with soft grip handles for comfort and control. The router has a lock-on button for continuous operation. It is ideal for trimming plastic laminates, wood veneers, and solid surface materials.