Learn More
Grainger Flush Trim Profile Router Bit 1/2
Catalog No. 502765079
Encompass_Preferred
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1.25 HP motor with a speed of 27,000 RPM. The router comes with a 1/4" collet and accepts 1/4" shank router bits. It features a depth stop for repeatable cuts and a dust extraction port to connect to a vacuum. The body is made of glass reinforced nylon for durability. This router is ideal for trimming edges of countertops, cabinets, furniture and more.
Catalog No. 50-276-5079 Supplier Grainger Supplier No. 16Y611
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1.25 HP motor with a speed of 27,000 RPM. The router comes with a 1/4" collet and accepts 1/4" shank router bits. It features a depth stop for repeatable cuts and a dust extraction port to connect to a vacuum. The body is made of glass reinforced nylon for durability. This router is ideal for trimming edges of countertops, cabinets, furniture and more.