Learn More
Grainger Flush Trim Profile Router Bit 1/4
Catalog No. 502765080
Encompass_Preferred
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 27,000 RPM. The router comes with a 1/4" collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It is lightweight at only 4.4 lbs. The router has a soft grip handle and trigger switch for comfort and control. It can be used for trimming countertop laminates, flooring, and wood veneers. This router helps provide smooth, professional results.
Catalog No. 50-276-5080 Supplier Grainger Supplier No. 16Y589
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 27,000 RPM. The router comes with a 1/4" collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It is lightweight at only 4.4 lbs. The router has a soft grip handle and trigger switch for comfort and control. It can be used for trimming countertop laminates, flooring, and wood veneers. This router helps provide smooth, professional results.