Learn More
Grainger Flush Trim Profile Router Bit 1/4
Catalog No. 502765227
Encompass_Preferred
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 27,000 RPM. The router comes with a 1/4" collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It is compact and lightweight at only 4 lbs. The soft grip handle provides comfort and control. This router allows for smooth trimming and profiling of countertop and cabinet edges.
Catalog No. 50-276-5227 Supplier Grainger Supplier No. 16Y561
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 27,000 RPM. The router comes with a 1/4" collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It is compact and lightweight at only 4 lbs. The soft grip handle provides comfort and control. This router allows for smooth trimming and profiling of countertop and cabinet edges.