Learn More
Grainger Flush Trim Profile Router Bit 1/2
Catalog No. 502767825
Encompass_Preferred
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 27,000 RPM. The router comes with a 1/4" collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It is lightweight at only 4.4 lbs. The router has a soft grip handle and trigger switch for comfort and control. Safety features include a spindle lock and double insulation. This router is ideal for trimming countertops, cabinets, and furniture edges.
Catalog No. 50-276-7825 Supplier Grainger Supplier No. 16Y546
The Westward Flush Trim Profile Router is a handheld power tool designed for trimming laminate and veneer edges. It has a 1-1/4 HP motor with a speed of 27,000 RPM. The router comes with a 1/4" collet, flush trim bit, edge guide, chip deflector, and dust extraction adapter. It is lightweight at only 4.4 lbs. The router has a soft grip handle and trigger switch for comfort and control. Safety features include a spindle lock and double insulation. This router is ideal for trimming countertops, cabinets, and furniture edges.