Learn More
Grainger Flush Trim Profile Router Bit 1/2
Catalog No. 502766860
Encompass_Preferred
The DeWalt DW6913 router bit set contains 15 high speed steel router bits in 1/4", 3/8", and 1/2" shank diameters. The set includes straight, roundover, cove, dovetail, chamfer, rabbeting, and flush trim router bits for use on wood, plastic, and non-ferrous metals. The bits have a titanium coating for increased durability and smooth cutting. The set comes in a plastic case for convenient storage and organization.
Catalog No. 50-276-6860 Supplier Grainger Supplier No. 16Y592
The DeWalt DW6913 router bit set contains 15 high speed steel router bits in 1/4", 3/8", and 1/2" shank diameters. The set includes straight, roundover, cove, dovetail, chamfer, rabbeting, and flush trim router bits for use on wood, plastic, and non-ferrous metals. The bits have a titanium coating for increased durability and smooth cutting. The set comes in a plastic case for convenient storage and organization.