Learn More
Ace Glass, Inc. FLUSH VALVE,30L+,AIR CONTROL
SDP
Catalog No. 5000854685
Encompass
Ace all PTFE reactor flush drain valve with control from your in-house air supply. Unique design weds our proven PTFE flush valve with a low profile air cylinder, control box and hoses. All wetted surfaces are PTFE or Kalrez. Valve will stay closed under heavy loads and temperature gradiants due to it's constant air supply. Loss of air pressure will not cause the valve to open. Order with your new reactor or replace an existing Ace flush drain valve.
Catalog No. 50-008-54685 Supplier Ace Glass, Inc. Supplier No. 1285410
Ace all PTFE reactor flush drain valve with control from your in-house air supply. Unique design weds our proven PTFE flush valve with a low profile air cylinder, control box and hoses. All wetted surfaces are PTFE or Kalrez. Valve will stay closed under heavy loads and temperature gradiants due to it's constant air supply. Loss of air pressure will not cause the valve to open. Order with your new reactor or replace an existing Ace flush drain valve.