Learn More
Med Vet International Flushing Kit w/ Spike and Bi-Directional V, J1339, Each
Supplier:  Med Vet International JOR1339
Encompass_Preferred
This kit allows controlled flushing. - Spike hooks to IV bag with flushing medication - Bivalve has female luerlock and male luerlock - Connect syringe to female luerlock to either flush or aspirate - Male luerlock: attach needle or catheter for flushing of ear, wound, joint, or other desired flush Vented IV spike with 36" (90cm) tubing connects to bivalve connector for flushing/aspiration. Sterile.
Catalog No. 50-005-81053