Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flusilazole | 85509-19-9 | MFCD01076615 | 97.7% | 315.39 | C16H15F2N3Si | 10 MG
SDP

Catalog No. 5000297466
Encompass_Preferred

Flusilazole analytical standard is an off-white solid reference material of the triazole fungicide flusilazole intended for analytical and research use. It is used as a reference in chromatographic and mass spectrometric method development, calibration, and quality-control applications.

  • Analytical standard for method development and quality control.
  • High purity (97.7%) for reliable analytical results.
  • CAS number 85509-19-9 for unambiguous identification.
  • Molecular weight 315.39 g/mol and formula C16H15F2N3Si.
  • Physical form: white to off-white solid.
  • Available in small pack sizes suitable for reference use (e.g., 10 mg).
  • Store under recommended conditions noted in the certificate of analysis.

Catalog No. 50-002-97466 Supplier Medchemexpress LLC Supplier No. HYB2012R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Flusilazole analytical standard is an off-white solid reference material of the triazole fungicide flusilazole intended for analytical and research use. It is used as a reference in chromatographic and mass spectrometric method development, calibration, and quality-control applications.

  • Analytical standard for method development and quality control.
  • High purity (97.7%) for reliable analytical results.
  • CAS number 85509-19-9 for unambiguous identification.
  • Molecular weight 315.39 g/mol and formula C16H15F2N3Si.
  • Physical form: white to off-white solid.
  • Available in small pack sizes suitable for reference use (e.g., 10 mg).
  • Store under recommended conditions noted in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.