Learn More
Medchemexpress LLC Flusilazole | 85509-19-9 | MFCD01076615 | 98.8% | 100 MG
SDP
Catalog No. 5000411070
Encompass_Preferred
Flusilazole is a broad-spectrum fungicide and cytochrome P-450 inhibitor used in studies related to fungal infections. It is intended for research use only.
  • Broad-spectrum fungicide
  • Cytochrome P-450 inhibitor
  • Used in studies related to fungal infections
  • Binds to cytochrome P-450 in *Saccharomyces cerevisiae* microsomes
  • Inhibits cholesterol 7α-hydroxylase activity in rat liver microsomes
  • Induces accumulation of lanosterol demethylase oxysterol intermediates
Catalog No. 50-004-11070 Supplier Medchemexpress LLC Supplier No. HYB2012100MG
Flusilazole is a broad-spectrum fungicide and cytochrome P-450 inhibitor used in studies related to fungal infections. It is intended for research use only.
  • Broad-spectrum fungicide
  • Cytochrome P-450 inhibitor
  • Used in studies related to fungal infections
  • Binds to cytochrome P-450 in *Saccharomyces cerevisiae* microsomes
  • Inhibits cholesterol 7α-hydroxylase activity in rat liver microsomes
  • Induces accumulation of lanosterol demethylase oxysterol intermediates