Learn More
Medchemexpress LLC HY-B1655 5mg Medchemexpress, Fluspirilene CAS:1841-19-6 Purity:>98%
SDP
Catalog No. 501932984
Encompass_Preferred
Medchemexpress, HY-B1655 5mg Fluspirilene CAS:1841-19-6 Fluspirilene is a non-competitive antagonist of L-type calcium channels with an IC 50 of 0.03 μM. Fluspirileneis a long-acting injectable depot antipsychotic drug used for schizophrenia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2984 Supplier Medchemexpress LLC Supplier No. HYB16555MG
Medchemexpress, HY-B1655 5mg Fluspirilene CAS:1841-19-6 Fluspirilene is a non-competitive antagonist of L-type calcium channels with an IC 50 of 0.03 μM. Fluspirileneis a long-acting injectable depot antipsychotic drug used for schizophrenia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.