Learn More
AdipoGen Flusulfamide
SDP
Supplier:  AdipoGen CDXF0082M100
Encompass_Preferred
Chemical. CAS 106917-52-6. Formula C13H7Cl2F3N2O4S. MW 415.2. Synthetic. Flusulfamide is primarily a soil-applied fungicide. It controls selected myxomycete fungi by preventing germination of both resting spores and secondary zoospores. The mode of action is not clear, however it activity might be attributed to cleavage of key structures within the spore cell walls. Compound can be used as analytical reference material.
Catalog No. 50-256-0890