Learn More
Apexbio Technology LLC Flutamide, 1g Cas# 13311-84-7
SDP
Supplier:  Apexbio Technology LLC B13761000
Encompass_Preferred
Flutamide is an antiandrogen drug, with its active metablolite binding at androgen receptor with K i values of 55 nM, and primarily used to treat prostate cancer. Phase 4. 13311-84-7. MFCD00072009 Flutamide is an antiandrogen drug, with its active metablolite binding at androgen receptor with Ki values of 55 nM, and primarily used to treat prostate cancer. Phase 4. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8611