Learn More
Selleck Chemical LLC Flutamide 100mg 13311-84-7 SCH-13521
SDP
Catalog No. 506622
Encompass_Preferred
Flutamide (SCH-13521) is an antiandrogen drug, with its active metablolite binding at androgen receptor with Ki values of 55 nM, and primarily used to treat prostate cancer. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-662-2 Supplier Selleck Chemical LLC Supplier No. S1908100MG
Flutamide (SCH-13521) is an antiandrogen drug, with its active metablolite binding at androgen receptor with Ki values of 55 nM, and primarily used to treat prostate cancer. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.