Learn More
Grainger Flute End Mill Set 10 Pcs
Catalog No. 502766915
Encompass_Preferred
The Dayton End Mill Set is a 2-piece set of 4-flute end mills made of cobalt steel with a TiAlN coating for wear resistance. The set includes one 1/4" diameter x 3/4" length end mill and one 1/2" diameter x 1-1/4" length end mill. Both end mills have a 1/2" shank diameter and are center cutting for plunge cuts. They are designed for high speed machining in aluminum, steel, stainless steel, titanium, and other materials. The end mills feature four flutes for efficient chip removal.
Catalog No. 50-276-6915 Supplier Grainger Supplier No. 16N203
The Dayton End Mill Set is a 2-piece set of 4-flute end mills made of cobalt steel with a TiAlN coating for wear resistance. The set includes one 1/4" diameter x 3/4" length end mill and one 1/2" diameter x 1-1/4" length end mill. Both end mills have a 1/2" shank diameter and are center cutting for plunge cuts. They are designed for high speed machining in aluminum, steel, stainless steel, titanium, and other materials. The end mills feature four flutes for efficient chip removal.