Learn More
Medchemexpress LLC Fluticasone | 90566-53-3 | 98.7% | 1 MG
SDP
Catalog No. 5000408305
Encompass_Preferred
Fluticasone is an inhaled corticosteroid utilized in respiratory research. It functions as a Smo agonist with an IC50 value of 99 nM. This compound is known to activate Hedgehog signaling and encourage the proliferation of primary neuronal stem or precursor cells.
  • Inhibits U2OS cell growth with an EC50 value of 99 nM
  • Decreases HRV-induced mucin production
  • Suppresses rhinovirus-induced airways inflammation in vivo
  • Activates Hedgehog signaling
  • Promotes proliferation of primary neuronal stem or precursor cells
Catalog No. 50-004-08305 Supplier Medchemexpress LLC Supplier No. HYB06031MG
Fluticasone is an inhaled corticosteroid utilized in respiratory research. It functions as a Smo agonist with an IC50 value of 99 nM. This compound is known to activate Hedgehog signaling and encourage the proliferation of primary neuronal stem or precursor cells.
  • Inhibits U2OS cell growth with an EC50 value of 99 nM
  • Decreases HRV-induced mucin production
  • Suppresses rhinovirus-induced airways inflammation in vivo
  • Activates Hedgehog signaling
  • Promotes proliferation of primary neuronal stem or precursor cells