Learn More
Medchemexpress LLC Fluticasone | 90566-53-3 | 98.7% | 1 ML
SDP
Catalog No. 5000408776
Encompass_Preferred
Fluticasone is an inhaled corticosteroid primarily used for respiratory research. It functions as a Smo agonist, exhibiting an IC50 value of 99 nM, and is known to activate Hedgehog signaling. This compound promotes the proliferation of primary neuronal stem or precursor cells.
  • Used for respiratory research
  • Acts as a Smo agonist with an IC50 of 99 nM
  • Activates Hedgehog signaling
  • Promotes proliferation of primary neuronal stem or precursor cells
  • Inhibits U2OS cell growth
  • Decreases HRV-induced mucin production
  • Suppresses rhinovirus-induced airways inflammation in vivo
Catalog No. 50-004-08776 Supplier Medchemexpress LLC Supplier No. HYB060310MM/1ML
Fluticasone is an inhaled corticosteroid primarily used for respiratory research. It functions as a Smo agonist, exhibiting an IC50 value of 99 nM, and is known to activate Hedgehog signaling. This compound promotes the proliferation of primary neuronal stem or precursor cells.
  • Used for respiratory research
  • Acts as a Smo agonist with an IC50 of 99 nM
  • Activates Hedgehog signaling
  • Promotes proliferation of primary neuronal stem or precursor cells
  • Inhibits U2OS cell growth
  • Decreases HRV-induced mucin production
  • Suppresses rhinovirus-induced airways inflammation in vivo