Learn More
Medchemexpress LLC HY-15234 10mg Medchemexpress, Fluticasone furoate CAS:397864-44-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1523410MG
Encompass_Preferred
Medchemexpress, HY-15234 10mg Fluticasone furoate CAS:397864-44-7 Fluticasone furoate is a topical, intranasal, enhanced-affinity synthetic trifluorinated corticosteroid with a Kd of 0.3 nM. Fluticasone furoate has potent anti-inflamatory and anti-asthmatic activity, and low systemic exposure. Fluticasone furoate has the potential for allergic rhinitis treatment[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0594