Learn More
Medchemexpress LLC Fluticasone furoate | 397864-44-7 | C27H29F3O6S | 1 ML
SDP
Catalog No. 5000372601
Encompass_Preferred
Fluticasone furoate is a topical, intranasal, enhanced-affinity synthetic trifluorinated corticosteroid with a Kd of 0.3 nM. It possesses potent anti-inflammatory and anti-asthmatic activity with low systemic exposure, making it suitable for allergic rhinitis treatment. This product is for research use only.
  • Potent anti-inflammatory and anti-asthmatic activity
  • Low systemic exposure
  • Suitable for allergic rhinitis treatment
  • Inhibits tumor necrosis factor synthesis and action
  • Prevents damage to cultured human lung epithelial cells
  • Primarily metabolized in the liver by CYP3A4
  • Mainly excreted in feces
Catalog No. 50-003-72601 Supplier Medchemexpress LLC Supplier No. HY1523410MM/1ML
Fluticasone furoate is a topical, intranasal, enhanced-affinity synthetic trifluorinated corticosteroid with a Kd of 0.3 nM. It possesses potent anti-inflammatory and anti-asthmatic activity with low systemic exposure, making it suitable for allergic rhinitis treatment. This product is for research use only.
  • Potent anti-inflammatory and anti-asthmatic activity
  • Low systemic exposure
  • Suitable for allergic rhinitis treatment
  • Inhibits tumor necrosis factor synthesis and action
  • Prevents damage to cultured human lung epithelial cells
  • Primarily metabolized in the liver by CYP3A4
  • Mainly excreted in feces