Learn More
Medchemexpress LLC Fluticasone furoate | 397864-44-7 | C27H29F3O6S | 100 MG
SDP
Catalog No. 5000373876
Encompass_Preferred
Fluticasone furoate is a topical, intranasal, enhanced-affinity synthetic trifluorinated corticosteroid with a Kd of 0.3 nM. It exhibits potent anti-inflammatory and anti-asthmatic activity with low systemic exposure. It has potential for allergic rhinitis treatment and effectively inhibits tumor necrosis factor synthesis and action.
  • Potent anti-inflammatory and anti-asthmatic activity
  • Low systemic exposure
  • Potential for allergic rhinitis treatment
  • Inhibits tumor necrosis factor synthesis and action
  • Prevents damage to cultured human lung epithelial cells from various stimuli
Catalog No. 50-003-73876 Supplier Medchemexpress LLC Supplier No. HY15234100MG
Fluticasone furoate is a topical, intranasal, enhanced-affinity synthetic trifluorinated corticosteroid with a Kd of 0.3 nM. It exhibits potent anti-inflammatory and anti-asthmatic activity with low systemic exposure. It has potential for allergic rhinitis treatment and effectively inhibits tumor necrosis factor synthesis and action.
  • Potent anti-inflammatory and anti-asthmatic activity
  • Low systemic exposure
  • Potential for allergic rhinitis treatment
  • Inhibits tumor necrosis factor synthesis and action
  • Prevents damage to cultured human lung epithelial cells from various stimuli