Learn More
Medchemexpress LLC Fluticasone furoate | 397864-44-7 | C27H29F3O6S | 500 MG
SDP
Catalog No. 5000373877
Encompass_Preferred
Fluticasone furoate is a topical, intranasal, enhanced-affinity synthetic trifluorinated corticosteroid with a Kd of 0.3 nM. It exhibits potent anti-inflammatory and anti-asthmatic activity with low systemic exposure and has potential for treating allergic rhinitis.
  • Potent anti-inflammatory and anti-asthmatic activity
  • Low systemic exposure
  • Potential for treating allergic rhinitis
  • Available as a nasal spray for topical administration
  • High potency in inhibiting tumor necrosis factor synthesis and action
  • Effective in preventing damage to cultured human lung epithelial cells
Catalog No. 50-003-73877 Supplier Medchemexpress LLC Supplier No. HY15234500MG
Fluticasone furoate is a topical, intranasal, enhanced-affinity synthetic trifluorinated corticosteroid with a Kd of 0.3 nM. It exhibits potent anti-inflammatory and anti-asthmatic activity with low systemic exposure and has potential for treating allergic rhinitis.
  • Potent anti-inflammatory and anti-asthmatic activity
  • Low systemic exposure
  • Potential for treating allergic rhinitis
  • Available as a nasal spray for topical administration
  • High potency in inhibiting tumor necrosis factor synthesis and action
  • Effective in preventing damage to cultured human lung epithelial cells