Learn More
Medchemexpress LLC Fluticasone propionate | 80474-14-2 | 99.9% | 5 MG
SDP
Catalog No. 5000406259
Encompass_Preferred
Fluticasone propionate is a potent topical anti-inflammatory corticosteroid and a selective glucocorticoid receptor agonist with an absolute affinity (KD) of 0.5 nM. It exhibits minimal or no activity at other steroid receptors and also possesses anti-viral activity.
  • Potent topical anti-inflammatory corticosteroid.
  • Selective glucocorticoid receptor agonist with a KD of 0.5 nM.
  • Exhibits minimal or no activity at other steroid receptors.
  • Possesses anti-viral activity.
  • Has an EC50 of 0.7 nM for transactivation activity at glucocorticoid receptor in HepG2 cells.
  • Shows an ED50 of 3 μg for anti-inflammatory activity in neutrophils.
  • Achieves an IC50 of 0.1 nM for inhibition of TNFalpha release in PBMC.
  • Demonstrates an EC50 of 1.09 nM for agonist activity at GR in SW1353 cells.
  • Inhibits TNFα-induced E-selectin expression with an IC50 of 1 nM.
Catalog No. 50-004-06259 Supplier Medchemexpress LLC Supplier No. HYB01545MG
Fluticasone propionate is a potent topical anti-inflammatory corticosteroid and a selective glucocorticoid receptor agonist with an absolute affinity (KD) of 0.5 nM. It exhibits minimal or no activity at other steroid receptors and also possesses anti-viral activity.
  • Potent topical anti-inflammatory corticosteroid.
  • Selective glucocorticoid receptor agonist with a KD of 0.5 nM.
  • Exhibits minimal or no activity at other steroid receptors.
  • Possesses anti-viral activity.
  • Has an EC50 of 0.7 nM for transactivation activity at glucocorticoid receptor in HepG2 cells.
  • Shows an ED50 of 3 μg for anti-inflammatory activity in neutrophils.
  • Achieves an IC50 of 0.1 nM for inhibition of TNFalpha release in PBMC.
  • Demonstrates an EC50 of 1.09 nM for agonist activity at GR in SW1353 cells.
  • Inhibits TNFα-induced E-selectin expression with an IC50 of 1 nM.