Learn More
Medchemexpress LLC Fluticasone (propionate) | 80474-14-2 | 99.9% | 50 MG
SDP
Catalog No. 5000405351
Encompass_Preferred
Fluticasone propionate is a potent topical anti-inflammatory corticosteroid that acts as a selective glucocorticoid receptor agonist with an absolute affinity (KD) of 0.5 nM. It exhibits minimal or no activity at other steroid receptors and possesses anti-viral activity. The product is for research use only.
  • Potent topical anti-inflammatory corticosteroid
  • Selective glucocorticoid receptor agonist
  • Absolute affinity (KD) of 0.5 nM
  • Little or no activity at other steroid receptors
  • Anti-viral activity
Catalog No. 50-004-05351 Supplier Medchemexpress LLC Supplier No. HYB015450MG
Fluticasone propionate is a potent topical anti-inflammatory corticosteroid that acts as a selective glucocorticoid receptor agonist with an absolute affinity (KD) of 0.5 nM. It exhibits minimal or no activity at other steroid receptors and possesses anti-viral activity. The product is for research use only.
  • Potent topical anti-inflammatory corticosteroid
  • Selective glucocorticoid receptor agonist
  • Absolute affinity (KD) of 0.5 nM
  • Little or no activity at other steroid receptors
  • Anti-viral activity