Learn More
Medchemexpress LLC Fluticasone propionate | 80474-14-2 | 99.93% | 200 MG
SDP
Catalog No. 5000406082
Encompass_Preferred
Fluticasone propionate is a potent topical anti-inflammatory corticosteroid and a selective glucocorticoid receptor agonist with an absolute affinity (KD) of 0.5 nM. It exhibits little to no activity at other steroid receptors and also possesses anti-viral activity. Its applications span immunology, anti-infection, and metabolic research.
  • Potent topical anti-inflammatory corticosteroid
  • Selective glucocorticoid receptor agonist (KD of 0.5 nM)
  • Exhibits minimal activity at other steroid receptors
  • Possesses anti-viral activity
  • Inhibits TNFα-induced E-selectin expression with an IC50 of 1 nM
  • Targets glucocorticoid receptor, endogenous metabolite, enterovirus, and ADC payload
  • Relevant to immunology/inflammation, vitamin D related/nuclear receptor, metabolic enzyme/protease, anti-infection, and antibody-drug conjugate/ADC related pathways
Catalog No. 50-004-06082 Supplier Medchemexpress LLC Supplier No. HYB0154200MG
Fluticasone propionate is a potent topical anti-inflammatory corticosteroid and a selective glucocorticoid receptor agonist with an absolute affinity (KD) of 0.5 nM. It exhibits little to no activity at other steroid receptors and also possesses anti-viral activity. Its applications span immunology, anti-infection, and metabolic research.
  • Potent topical anti-inflammatory corticosteroid
  • Selective glucocorticoid receptor agonist (KD of 0.5 nM)
  • Exhibits minimal activity at other steroid receptors
  • Possesses anti-viral activity
  • Inhibits TNFα-induced E-selectin expression with an IC50 of 1 nM
  • Targets glucocorticoid receptor, endogenous metabolite, enterovirus, and ADC payload
  • Relevant to immunology/inflammation, vitamin D related/nuclear receptor, metabolic enzyme/protease, anti-infection, and antibody-drug conjugate/ADC related pathways