Learn More
Medchemexpress LLC Fluticasone propionate | 80474-14-2 | 99.93% | 500 MG
SDP
Catalog No. 5000406237
Encompass_Preferred
Fluticasone propionate is a potent topical anti-inflammatory corticosteroid. It acts as a selective glucocorticoid receptor agonist, exhibiting an absolute affinity (KD) of 0.5 nM. The product shows minimal to no activity at other steroid receptors and also possesses anti-viral activity.
  • Potent topical anti-inflammatory corticosteroid
  • Selective glucocorticoid receptor agonist
  • Absolute affinity (KD) of 0.5 nM
  • Minimal to no activity at other steroid receptors
  • Possesses anti-viral activity
Catalog No. 50-004-06237 Supplier Medchemexpress LLC Supplier No. HYB0154500MG
Fluticasone propionate is a potent topical anti-inflammatory corticosteroid. It acts as a selective glucocorticoid receptor agonist, exhibiting an absolute affinity (KD) of 0.5 nM. The product shows minimal to no activity at other steroid receptors and also possesses anti-viral activity.
  • Potent topical anti-inflammatory corticosteroid
  • Selective glucocorticoid receptor agonist
  • Absolute affinity (KD) of 0.5 nM
  • Minimal to no activity at other steroid receptors
  • Possesses anti-viral activity