Learn More
Medchemexpress LLC Fluticasone propionate-d5 | 1093258-28-6 | 91.7% | 1 MG
SDP
Catalog No. 5000407240
Encompass_Preferred
Fluticasone propionate-d5 is a deuterium labeled form of Fluticasone propionate. It is a potent topical anti-inflammatory corticosteroid and a selective glucocorticoid receptor agonist with an absolute affinity (KD) of 0.5 nM. It exhibits little to no activity at other steroid receptors and possesses anti-viral activity.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07240 Supplier Medchemexpress LLC Supplier No. HYB0154S11MG
Fluticasone propionate-d5 is a deuterium labeled form of Fluticasone propionate. It is a potent topical anti-inflammatory corticosteroid and a selective glucocorticoid receptor agonist with an absolute affinity (KD) of 0.5 nM. It exhibits little to no activity at other steroid receptors and possesses anti-viral activity.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.