Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluticasone Propionate, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18602796 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-796 25 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-602-796 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F301225GMBL

F3012, 80474-14-2, C25H31F3O5S

Spectrum™ Chemical Fluticasone Propionate, USP is used to prevent symptoms of asthma, treat the symptoms of allergic and non-allergic rhinitis, and relieve inflammation and itching caused by various skin conditions. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 80474-14-2
Molecular Formula C25H31F3O5S
Molecular Weight (g/mol) 500.57
InChI Key WMWTYOKRWGGJOA-ZHLGSTKJSA-N
IUPAC Name (1R,2R,3aS,5S,9aS,9bR,10S,11aS)-5,9b-difluoro-1-{[(fluoromethyl)sulfanyl]carbonyl}-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl propanoate
SMILES CCC(=O)O[C@@]1([C@H](C)C[C@H]2C3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SCF

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 25 g
Specific Rotation +32° to +36°
Percent Purity 98 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.