Learn More
Medchemexpress LLC Flutrimazole | 119006-77-8 | 99.3% | 10 MG
SDP
Catalog No. 5000339508
Encompass_Preferred
Flutrimazole is an imidazole antifungal with dual anti-inflammatory and antifungal activity. It shows scarce transdermal penetration and is advantageous in the research of topical fungal infections with an inflammatory component. This product is for research use only and not sold to patients.
  • Imidazole antifungal with dual anti-inflammatory and antifungal activity
  • Scarce transdermal penetration
  • Advantageous in the research of topical fungal infections with an inflammatory component
  • For research use only
Catalog No. 50-003-39508 Supplier Medchemexpress LLC Supplier No. HY12906010MG
Flutrimazole is an imidazole antifungal with dual anti-inflammatory and antifungal activity. It shows scarce transdermal penetration and is advantageous in the research of topical fungal infections with an inflammatory component. This product is for research use only and not sold to patients.
  • Imidazole antifungal with dual anti-inflammatory and antifungal activity
  • Scarce transdermal penetration
  • Advantageous in the research of topical fungal infections with an inflammatory component
  • For research use only