Learn More
Medchemexpress LLC Flutrimazole | 119006-77-8 | 99.3% | 50 MG
SDP
Catalog No. 5000340018
Encompass_Preferred
Flutrimazole is an imidazole antifungal with dual anti-inflammatory and antifungal activity. It shows scarce transdermal penetration and is advantageous in the research of topical fungal infections with an inflammatory component.
  • Imidazole antifungal
  • Dual anti-inflammatory and antifungal activity
  • Scarce transdermal penetration
  • Suitable for research into topical fungal infections with an inflammatory component
Catalog No. 50-003-40018 Supplier Medchemexpress LLC Supplier No. HY12906050MG
Flutrimazole is an imidazole antifungal with dual anti-inflammatory and antifungal activity. It shows scarce transdermal penetration and is advantageous in the research of topical fungal infections with an inflammatory component.
  • Imidazole antifungal
  • Dual anti-inflammatory and antifungal activity
  • Scarce transdermal penetration
  • Suitable for research into topical fungal infections with an inflammatory component