Learn More
Medchemexpress LLC Flutrimazole | 119006-77-8 | MFCD00865591 | 99.3% | 346.37 | C22H16F2N2 | 5 MG
SDP
Catalog No. 5000262729
Encompass_Preferred
Flutrimazole is an imidazole antifungal analytical standard (CAS 119006-77-8) used in research and analytical applications. It shows dual anti-inflammatory and antifungal activity, limited transdermal penetration, and is supplied as a high-purity solid suitable for chromatographic and mass spectrometry methods.
  • High purity (99.3%) for reliable analytical results.
  • Suitable for HPLC, GC, and MS method development and validation.
  • Dual anti-inflammatory and antifungal profile useful for topical infection studies.
  • Limited transdermal penetration reduces systemic absorption concerns in dermal models.
  • Available in small sample sizes for analytical testing, such as 5 mg.
Catalog No. 50-002-62729 Supplier Medchemexpress LLC Supplier No. HY1290605MG
Flutrimazole is an imidazole antifungal analytical standard (CAS 119006-77-8) used in research and analytical applications. It shows dual anti-inflammatory and antifungal activity, limited transdermal penetration, and is supplied as a high-purity solid suitable for chromatographic and mass spectrometry methods.
  • High purity (99.3%) for reliable analytical results.
  • Suitable for HPLC, GC, and MS method development and validation.
  • Dual anti-inflammatory and antifungal profile useful for topical infection studies.
  • Limited transdermal penetration reduces systemic absorption concerns in dermal models.
  • Available in small sample sizes for analytical testing, such as 5 mg.