Learn More
FUJIFILM Biosciences Fluvalinate Standard
Catalog No. 5000450405
Encompass_Preferred
Fluvalinate Standard is a high-purity reference material 97 0% by HPLC used for pesticide residue analysis Designed for use in environmental and food safety testing This standard supports accurate calibration in HPLC methods for pyrethroid pesticide quantification Clear colorless to pale yellowish brown liquid
Catalog No. 50-004-50405 Supplier FUJIFILM Biosciences Supplier No. 06102471
Fluvalinate Standard is a high-purity reference material 97 0% by HPLC used for pesticide residue analysis Designed for use in environmental and food safety testing This standard supports accurate calibration in HPLC methods for pyrethroid pesticide quantification Clear colorless to pale yellowish brown liquid