Learn More
Medchemexpress LLC Fluvastatin-d6 sodium | 00-00-0 | 98.1% | C24H19D6FNNaO4 | 10MG
SDP
Catalog No. 5000259556
Encompass_Preferred
Fluvastatin-d6 sodium is a deuterium-labeled research standard of fluvastatin sodium, intended for use as an internal standard or tracer in mass spectrometry, pharmacokinetic, and metabolic studies. It preserves the biochemical profile of the parent HMG-CoA reductase inhibitor and is supplied at high purity for quantitative and qualitative analyses; for research use only.
  • Deuterium-labeled analog for accurate mass spectrometry quantitation.
  • High purity (98.1%) for reliable analytical results.
  • Molecular formula C24H19D6FNNaO4 supports identity confirmation.
  • Suitable as an internal standard in pharmacokinetic and metabolic studies.
  • Available in small pack sizes suitable for analytical workflows.
  • Supplied for research use only; not for human or clinical use.
Catalog No. 50-002-59556 Supplier Medchemexpress LLC Supplier No. HY14664AS10MG
Fluvastatin-d6 sodium is a deuterium-labeled research standard of fluvastatin sodium, intended for use as an internal standard or tracer in mass spectrometry, pharmacokinetic, and metabolic studies. It preserves the biochemical profile of the parent HMG-CoA reductase inhibitor and is supplied at high purity for quantitative and qualitative analyses; for research use only.
  • Deuterium-labeled analog for accurate mass spectrometry quantitation.
  • High purity (98.1%) for reliable analytical results.
  • Molecular formula C24H19D6FNNaO4 supports identity confirmation.
  • Suitable as an internal standard in pharmacokinetic and metabolic studies.
  • Available in small pack sizes suitable for analytical workflows.
  • Supplied for research use only; not for human or clinical use.