Learn More
Medchemexpress LLC Fluvastatin-d6 sodium (XU 62-320-d6) | 93957-55-2 | 98.06% | 439.48 | 1 MG
SDP
Catalog No. 5000362977
Encompass_Preferred
Fluvastatin-d6 (sodium) is deuterium labeled Fluvastatin sodium. Fluvastatin sodium (XU 62320) is a first fully synthetic, competitive HMG-CoA reductase inhibitor with an IC50 of 8 nM. It is primarily intended for research applications.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-62977 Supplier Medchemexpress LLC Supplier No. HY14664AS1MG
Fluvastatin-d6 (sodium) is deuterium labeled Fluvastatin sodium. Fluvastatin sodium (XU 62320) is a first fully synthetic, competitive HMG-CoA reductase inhibitor with an IC50 of 8 nM. It is primarily intended for research applications.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.