Learn More
Apexbio Technology LLC Fluvastatin Sodium 93957-55-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061869
Encompass_Preferred
Fluvastatin Sodium (CAS 93957-55-2) is an inhibitor of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase the rate-limiting enzyme in cholesterol biosynthesis By blocking HMG-CoA reductase activity fluvastatin reduces serum total cholesterol LDL-cholesterol and apolipoprotein B while increasing HDL-cholesterol and apolipoprotein A-I levels In research settings fluvastatin exhibits anti-atherosclerotic anti-thrombotic and antioxidant properties Studies have shown fluvastatin decreases platelet aggregation dose-dependently in vitro reducing platelet aggregation by approximately 10-15% at clinical doses (40 mg/day) Additionally fluvastatin inhibits inflammatory angiogenesis and NO production suggesting potential applications in inflammatory and cardiovascular research
Catalog No. 50-000-61869 Supplier Apexbio Technology LLC Supplier No. A43635.1
Fluvastatin Sodium (CAS 93957-55-2) is an inhibitor of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase the rate-limiting enzyme in cholesterol biosynthesis By blocking HMG-CoA reductase activity fluvastatin reduces serum total cholesterol LDL-cholesterol and apolipoprotein B while increasing HDL-cholesterol and apolipoprotein A-I levels In research settings fluvastatin exhibits anti-atherosclerotic anti-thrombotic and antioxidant properties Studies have shown fluvastatin decreases platelet aggregation dose-dependently in vitro reducing platelet aggregation by approximately 10-15% at clinical doses (40 mg/day) Additionally fluvastatin inhibits inflammatory angiogenesis and NO production suggesting potential applications in inflammatory and cardiovascular research