Learn More
TARGETMOL CHEMICALS INC Fluvastatin sodium 200MG
SDP
Catalog No. 502878292
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Fluvastatin sodium (Fluvastatin sodium salt), a competitive inhibitor of hydroxymethylglutaryl-coenzyme A reductase (HMGCR), is a commonly used cholesterol lowering agent. Purity 99.46%
Catalog No. 50-287-8292 Supplier TARGETMOL CHEMICALS INC Supplier No. T1487200MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Fluvastatin sodium (Fluvastatin sodium salt), a competitive inhibitor of hydroxymethylglutaryl-coenzyme A reductase (HMGCR), is a commonly used cholesterol lowering agent. Purity 99.46%