Learn More
Apexbio Technology LLC Fluvastatin Sodium, 50mg. Cas: 93957-55-2 MFCD: MFCD00929076. HMG-CoA reductase inhibitor
SDP
Catalog No. 501904929
Encompass_Preferred
Fluvastatin sodium is an inhibitor of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase [1].Fluvastatin is a drug used to treat patients with hypercholesterolaemia. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4929 Supplier Apexbio Technology LLC Supplier No. A436350
Fluvastatin sodium is an inhibitor of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase [1].Fluvastatin is a drug used to treat patients with hypercholesterolaemia. Other sizes are available. Please inqury us for quote.