Learn More
Medchemexpress LLC Fluvastatin sodium | 94061-80-0 | C24H25FNNaO4 | 10MG
SDP
Catalog No. 5000259443
Encompass_Preferred
3R,5S-Fluvastatin sodium is the monosodium salt of the (3R,5S) enantiomer of fluvastatin, a synthetic HMG-CoA reductase inhibitor. It is provided as a research-grade reagent and analytical standard for biochemical and pharmacological studies of cholesterol biosynthesis inhibition.
  • High purity (~99.2%) for reproducible results.
  • Monosodium salt form improves water solubility.
  • Suitable for enantiomer-specific biochemical and pharmacology studies.
  • Characterized: C24H25FNNaO4, molecular weight 433.4 g/mol.
  • Supplied with lot-specific certificate of analysis for traceability.
Catalog No. 50-002-59443 Supplier Medchemexpress LLC Supplier No. HY14664F10MG
3R,5S-Fluvastatin sodium is the monosodium salt of the (3R,5S) enantiomer of fluvastatin, a synthetic HMG-CoA reductase inhibitor. It is provided as a research-grade reagent and analytical standard for biochemical and pharmacological studies of cholesterol biosynthesis inhibition.
  • High purity (~99.2%) for reproducible results.
  • Monosodium salt form improves water solubility.
  • Suitable for enantiomer-specific biochemical and pharmacology studies.
  • Characterized: C24H25FNNaO4, molecular weight 433.4 g/mol.
  • Supplied with lot-specific certificate of analysis for traceability.