Learn More
Enzo Life Sciences Fluvastatin. sodium salt (10mg). CAS: 93957-55-2
SDP
Supplier:  Enzo Life Sciences ALX270466M010
Encompass_Preferred
Anticholesterol agent. HMG-CoA reductase inhibitor and antioxidant. Purity: ≥98% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8589