Learn More
Abcam Fluvastatin sodium salt, HMG-CoA reductase inhibitor, 50MG
Catalog No. 502707386
Encompass_Preferred
MW 433.45 g/mol, Purity >99%. Competitive HMG-CoA reductase inhibitor (IC₅₀ = 40 - 100 nM in human and rat microsomes). Potent inhibitor of vascular smooth muscle cell proliferation in vitro (IC₅₀ = 70 nM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7386 Supplier Abcam Supplier No. AB12065150MG
MW 433.45 g/mol, Purity >99%. Competitive HMG-CoA reductase inhibitor (IC₅₀ = 40 - 100 nM in human and rat microsomes). Potent inhibitor of vascular smooth muscle cell proliferation in vitro (IC₅₀ = 70 nM).
The product is subject to the following: Abcam Restricted Use Statement