Learn More
Selleck Chemical LLC Fluvastatin Sodium 50mg 93957-55-2 XU-62-320
SDP
Supplier:  Selleck Chemical LLC S190950MG
Encompass_Preferred
Fluvastatin Sodium (XU-62-320) inhibits HMG-CoA reductase activity with IC50 of 8 nM in a cell-free assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-555-2