Learn More
Cayman Chemical FluvoxamIn-d4maleate 1mg
SDP
Catalog No. 502911456
Encompass_Preferred
An internal standard for the quantification of fluvoxamine by GC- or LC-MS
Catalog No. 50-291-1456 Supplier Cayman Chemical Supplier No. 348851 MG
Add to Cart
Add to Cart
An internal standard for the quantification of fluvoxamine by GC- or LC-MS