Learn More
Cayman Chemical FluvoxamIn-d4maleate 500ug
SDP
Catalog No. 502911450
Encompass_Preferred
An internal standard for the quantification of fluvoxamine by GC- or LC-MS
Catalog No. 50-291-1450 Supplier Cayman Chemical Supplier No. 34885500 UG
Add to Cart
Add to Cart
An internal standard for the quantification of fluvoxamine by GC- or LC-MS