Learn More
Cayman Chemical FluvoxamIn-d4maleate 5mg
SDP
Catalog No. 502910524
Encompass_Preferred
An internal standard for the quantification of fluvoxamine by GC- or LC-MS
Catalog No. 50-291-0524 Supplier Cayman Chemical Supplier No. 348855 MG
An internal standard for the quantification of fluvoxamine by GC- or LC-MS