Learn More
Selleck Chemical LLC Fluvoxamine (DU-23000) maleate S1336-50mg
SDP
Catalog No. 503111361
Encompass_Preferred
Fluvoxamine maleate (DU-23000 MK-264) is a selective serotonin (5-HT) reuptake inhibitor (SSRI) used in the treatment of obsessive-compulsive disorder
Catalog No. 50-311-1361 Supplier Selleck Chemical LLC Supplier No. S133650MG
Fluvoxamine maleate (DU-23000 MK-264) is a selective serotonin (5-HT) reuptake inhibitor (SSRI) used in the treatment of obsessive-compulsive disorder