Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluvoxamine Maleate 98.0+%, TCI America™
SDP

Catalog No. F08585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08585G 5 g
F08581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08585G Supplier TCI America Supplier No. F08585G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 61718-82-9
Molecular Formula C19H25F3N2O6
Molecular Weight (g/mol) 434.41
MDL Number MFCD00269809,MFCD00269809
InChI Key LFMYNZPAVPMEGP-PIDGMYBPSA-N
Synonym (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone O-(2-Aminoethyl)oxime Maleate
PubChem CID 119090957
IUPAC Name (2Z)-but-2-enedioic acid; (E)-(2-aminoethoxy)({5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene})amine
SMILES OC(=O)\C=C/C(O)=O.COCCCC\C(=N/OCCN)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 122°C
Color White
Quantity 5 g
Formula Weight 434.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fluvoxamine Maleate
RTECSNumber SA9230000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.